missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Inoculating Nichrome Loops 8 in. Handle

Catalog No. 13820030
Change view
Click to view available options
Size:
2 mm
3 mm
4 mm
5 mm
Includes:
Nichrome wire with aluminum handle
Nichrome wire with insulated handle
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Includes
13820030 5 mm Nichrome wire with aluminum handle
13820027 2 mm Nichrome wire with aluminum handle
13820028 3 mm Nichrome wire with aluminum handle
13820029 4 mm Nichrome wire with aluminum handle
13820031 2 mm Nichrome wire with insulated handle
13820032 3 mm Nichrome wire with insulated handle
13820033 4 mm Nichrome wire with insulated handle
13820034 5 mm Nichrome wire with insulated handle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 13820030 Supplier Fisherbrand™ Supplier No. 10401131

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Fisherbrand™ Innoculating loops are available in a variety of gauges, loop hole sizes, and wire types to accommodate an array of laboratory requirements.

Fisherbrand™ Nichrome/Aluminum Innoculating Loops consist of a nichrome wire loop mounted to an aluminum handle. The aluminum handle is available in either an insulated or non-insulated format.

  • Ideal for selecting, inoculating, or transferring microbiological samples
  • Cools rapidly after sterilization
  • Loops Slide along agar surfaces without cutting or tearing material
  • Nichrome wire available in single or twisted configuration
  • Passivated
  • 203 mm (8 in.) handles, available with insulation
  • Compatible with Gamma/ETO/Autoclavable sterilization methods

Certifications

ISO 9001-02, GMP, cGMP

Specifications

Material Nichrome
Length (English) 11 in.
Length (Metric) 280 mm
Size 5 mm
Color Silver
Product Type Inoculating Loop
For Use With (Equipment) Petri-Dish etc
Gauge 25
Includes Nichrome wire with aluminum handle
Quantity Available as Each or Pk. of 12
Autoclavable Autoclavable
For Use With (Application) Ideal for selecting, inoculating, or transferring microbiological samples
Show More Show Less