missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Inoculating Nichrome Loops 8 in. Handle
Fisherbrand™ Innoculating loops are available in a variety of gauges, loop hole sizes, and wire types to accommodate an array of laboratory requirements.
Supplier: Fisherbrand™ 10401130
Description
Fisherbrand™ Nichrome/Aluminum Innoculating Loops consist of a nichrome wire loop mounted to an aluminum handle. The aluminum handle is available in either an insulated or non-insulated format.
- Ideal for selecting, inoculating, or transferring microbiological samples
- Cools rapidly after sterilization
- Loops Slide along agar surfaces without cutting or tearing material
- Nichrome wire available in single or twisted configuration
- Passivated
- 203 mm (8 in.) handles, available with insulation
- Compatible with Gamma/ETO/Autoclavable sterilization methods
Certifications
ISO 9001-02, GMP, cGMP
Chemical Identifiers
| 11 in. | |
| Nichrome | |
| Inoculating Loop | |
| Available as Each or Pk. of 12 | |
| Ideal for selecting, inoculating, or transferring microbiological samples |
| 280 mm | |
| Silver | |
| Petri-Dish etc | |
| Autoclavable |
Specifications
| 11 in. | |
| Nichrome | |
| Silver | |
| Petri-Dish etc | |
| Nichrome wire with aluminum handle | |
| Autoclavable |
| 280 mm | |
| 4 mm | |
| Inoculating Loop | |
| 25 | |
| Available as Each or Pk. of 12 | |
| Ideal for selecting, inoculating, or transferring microbiological samples |