missing translation for 'onlineSavingsMsg'
Learn More

Innovatek Innovacon Strep A Rapid Test Kits

Catalog No. 361045031
Change view
Click to view available options
Quantity:
25 Tests/Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
361045031 25 Tests/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 361045031 Supplier Innovatek Supplier No. 4010IST501
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Innovacon Strep A Rapid Test Kits provide results within 5 minutes

  • Rapid detection of Group A Streptococcus bacterial antigen
  • High sensitivity at low colony count
  • Accurate results directly from throat swab
  • Results in 5 minutes
  • Easy to read control and test lines
  • All materials included in the kit

Specifications

Detectable Analytes Streptococci (Group A)
Format Throat Swab
Sensitivity Highly Sensitive at Low Colony Counts
Disposable Single-use
For Use With (Application) Strep A Detection
Quantity 25 Tests/Kit
Test Time 5 min.