missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Synthware™ Inlet Feed Tubes
For use with rotary evaporator condensers
$283.00 - $283.00
Specifications
| Material | Borosilicate Glass |
|---|---|
| For Use With (Application) | Organic Synthesis |
| Color | Clear |
| Product Type | Inlet Feed Tube |