missing translation for 'onlineSavingsMsg'
Learn More

METTLER TOLEDO™ InLab™ Surface pH Electrodes

Catalog No. 01911474
Change view
Click to view available options
Connector Types:
Multi-Pin
S7
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connector Types
01911474 S7
01914309 Multi-Pin
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01911474 Supplier METTLER TOLEDO™ Supplier No. 51343157
Only null left

A pH electrode featuring a flat, rugged plastic membrane for measuring samples with flat surfaces.

  • For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
  • Intelligent Sensor Management (ISM) electrodes capture and save calibration data, which is then automatically recognized by the meter once connected.
  • Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
  • Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.

Featured Applications

  • Agar plates
  • Drop size samples
  • Paper
  • Skin
  • Leather
  • Suspended solids
  • Textiles and Prints

Accessories & Care

  • Ceramic ring junction
  • Polysulfone membrane
  • Refillable 3M KCl (51350072): Keep probe filled with electrolyte for accurate results
  • Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
  • Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)

Specifications

Connector Types S7
Junction Ceramic
Material Polysulfone
pH Range 1 to 11
Temperature (Metric) Operating 0°C to 50°C
Type Combination Electrode
Filling Solution Type Saturated AgCl
For Use With (Application) For measuring samples such as Skin, Leather, Paper and Agar
For Use With (Equipment) S7 to BNC (30281915)
Includes Flat membrane
Internal Reference Ag/AgCl
Membrane LoT Glass
Model InLab™ Surface
Response Time <20 sec.
Storage Requirements 3mol/L KCl
Temperature (English) Operating 32°F to 122°F
Temperature (Metric) Range 0°C to 50°C
Length (English) 4.33 in.
Length (Metric) 110 mm
Diameter (English) 0.47 in.
Diameter (Metric) 12 mm
Show More Show Less