missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mettler Toledo™ InLab™ Sensor Cables: S7 Connection
Electrode cables to connect Mettler Toledo InLab sensors to any Mettler Toledo meter.
$132.50 - $216.37
Specifications
| For Use With (Equipment) | Mettler Toledo InLab Sensors |
|---|---|
| For Use With (Application) | To connect Mettler Toledo InLab sensors to any meter |
| Catalog Number | Mfr. No. | Connector Types | Length (Metric) Cable | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Connector Types | Length (Metric) Cable | Price | Quantity & Availability | ||||
|
01914282
|
Mettler Toledo™
30281916 |
S7-BNC | 3.0 m |
Each for $216.37
|
|
||||
|
01914281
|
Mettler Toledo™
30281915 |
S7-BNC | 1.2 m |
Each for $132.50
|
|
||||
Description
- Detachable
- Resuable
Specifications
| Mettler Toledo InLab Sensors | |
| To connect Mettler Toledo InLab sensors to any meter |