missing translation for 'onlineSavingsMsg'
Learn More
Learn More
METTLER TOLEDO™ InLab™ Routine pH Electrodes
Description
- For use with METTLER TOLEDO SevenExcellence, SevenCompact and portable meters as well as other brands. Connection cables may be required.
- Intelligent Sensor Management (ISM) electrodes capture and save calibration data, which is then automatically recognized by the meter once connected.
- Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
- Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.
Featured Applications
- Drinking water
- Highly saline solutions or seawater
- Microbiological samples
- Corrosive acids and bases
- General Lab
- Chemical
- High pH values
- TRIS compatible
Accessories & Care
- Refillable 3M KCl (51350072): Keep probe filled with electrolyte for accurate results
- Tip: Open fill port while measuring, and also to refill electrolyte solution
- Ceramic junction
- Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
- Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
Order Info
Cable not included (sold separately).
Recommended Storage
InLab storage solution
Specifications
Specifications
| Description | pH Electrode InLab Routine Go-ISM |
| Connector Types | BNC IP67 |
| Junction | Ceramic |
| Material | Glass |
| pH Range | 0 to 14 |
| Temperature (Metric) Operating | 0°C to 100°C |
| Type | Combination Electrode |
| For Use With (Application) | Mobile applications |
| For Use With (Equipment) | Fixed Cable to sensor |
| Includes | ISM Temperature Probe |
| Show More |