missing translation for 'onlineSavingsMsg'
Learn More

METTLER TOLEDO™ InLab™ Routine pH Electrodes

A combination pH electrode with an integrated temperature sensor and chemically resistant HA glass body. It is designed for general lab applications and ideal for use at high pH values (IP67 rated).

Supplier:  METTLER TOLEDO™ 51343056

 View more versions of this product

Catalog No. 01911456


Only null left

Description

Description

  • For use with METTLER TOLEDO SevenExcellence, SevenCompact and portable meters as well as other brands. Connection cables may be required.
  • Intelligent Sensor Management (ISM) electrodes capture and save calibration data, which is then automatically recognized by the meter once connected.
  • Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
  • Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.

Featured Applications

  • Drinking water
  • Highly saline solutions or seawater
  • Microbiological samples
  • Corrosive acids and bases
  • General Lab
  • Chemical
  • High pH values
  • TRIS compatible

Accessories & Care

  • Refillable 3M KCl (51350072): Keep probe filled with electrolyte for accurate results
  • Tip: Open fill port while measuring, and also to refill electrolyte solution
  • Ceramic junction
  • Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
  • Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)

Order Info

Cable not included (sold separately).

Specifications

Chemical Identifiers

Ceramic
0 to 14
Combination Electrode
0°C to 100°C
Glass
0°C to 100°C
HA Glass

Specifications

Multi-Pin
Glass
0°C to 100°C
3 mol/L KCl
Multi-Pin to BNC (30281896)
ARGENTHAL
InLab™ Routine Pt1000
<20 sec.
32°F to 212°F
4.72 in.
0.47 in.
Ceramic
0 to 14
Combination Electrode
Accurate measurement in a wide range of sample types
Temperature Probe (Pt1000)
HA Glass
PT1000 ATC
3 mol/L KCl
0°C to 100°C
120 mm
12 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions