missing translation for 'onlineSavingsMsg'
Learn More

METTLER TOLEDO™ InLab™ Routine pH Electrodes

Catalog No. 01911454
Change view
Click to view available options
Connector Types:
BNC IP67
Multi-Pin
S7
Includes:
Electrode only
ISM Temperature Probe
Temperature Probe
Temperature Probe (Pt1000)
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connector Types Includes
01911454 Multi-Pin Temperature Probe
01911450 S7 Electrode only
01911456 Multi-Pin Temperature Probe (Pt1000)
01917142 Multi-Pin ISM Temperature Probe
01914301 BNC IP67 ISM Temperature Probe
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01911454 Supplier METTLER TOLEDO™ Supplier No. 51343054

A combination pH electrode with an integrated temperature sensor and chemically resistant HA glass body. It is designed for general lab applications and ideal for use at high pH values (IP67 rated).

  • For use with METTLER TOLEDO SevenExcellence, SevenCompact and portable meters as well as other brands. Connection cables may be required.
  • Intelligent Sensor Management (ISM) electrodes capture and save calibration data, which is then automatically recognized by the meter once connected.
  • Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
  • Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.

Featured Applications

  • Drinking water
  • Highly saline solutions or seawater
  • Microbiological samples
  • Corrosive acids and bases
  • General Lab
  • Chemical
  • High pH values
  • TRIS compatible

Accessories & Care

  • Refillable 3M KCl (51350072): Keep probe filled with electrolyte for accurate results
  • Tip: Open fill port while measuring, and also to refill electrolyte solution
  • Ceramic junction
  • Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
  • Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)

Order Info

Cable not included (sold separately).

Specifications

Connector Types Multi-Pin
Junction Ceramic
Material Glass
pH Range 0 to 14
Temperature (Metric) Operating 0°C to 100°C
Type Combination Electrode
Filling Solution Type 3 mol/L KCl
For Use With (Application) Accurate measurement in a wide range of sample types
For Use With (Equipment) Multi-Pin to BNC (30281896)
Includes Temperature Probe
Internal Reference ARGENTHAL
Membrane HA Glass
Model InLab™ Routine Pro
Probe NTC 30 kilohm ATC
Response Time <20 sec.
Storage Requirements 3 mol/L KCl
Temperature (English) Operating 32°F to 212°F
Temperature (Metric) Range 0°C to 100°C
Length (English) 4.72 in.
Length (Metric) 120 mm
Diameter (English) 0.47 in.
Diameter (Metric) 12 mm
Show More Show Less