missing translation for 'onlineSavingsMsg'
Learn More
Learn More
METTLER TOLEDO™ InLab™ Power pH Electrodes
Description
- For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
- Intelligent Sensor Management (ISM) electrodes capture and save calibration data, which is then automatically recognized by the meter once connected.
- Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
- Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.
Featured Applications
- Corrosive acids and bases
- Protein-rich samples
- Dairy
- High pH values
- High temperature
- Junction-clogging samples
- Polymeric dispersions
- TRIS compatible
Accessories & Care
- Non-refillable - no additional electrolyte required
- Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
- Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
Specifications
Specifications
| Description | pH Electrode InLab Power |
| Connector Types | S7 |
| Junction | Ceramic |
| Material | Glass |
| pH Range | 0 to 12 |
| Temperature (Metric) Operating | 0°C to 130°C |
| Type | Combination Electrode |
| Filling Solution Type | DPA Gel |
| For Use With (Application) | High temperatures and biological media |
| For Use With (Equipment) | S7 to BNC (30281915) |
| Show More |