missing translation for 'onlineSavingsMsg'
Learn More
Learn More
METTLER TOLEDO™ InLab™ Max Pro-ISM pH Electrode
Description
- For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
- Intelligent Sensor Management (ISM) electrodes capture and save calibration data, which is then automatically recognized by the meter once connected.
- Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
- Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.
Featured Applications
- High-ion strength
- High temperature samples
- Wastewater
- TRIS buffers
- Pharmaceutical & Biological samples
- Organic solvents
- Fruit juice
- Water
- General lab
Accessories & Care
- Immovable glass sleeve junction
- Refillable 3M KCl (51350072) - Keep probe filled with electrolyte for accurate results
- Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
- Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
Recommended Storage
InLab storage solution
Specifications
Specifications
| Connector Types | Multi-Pin |
| Junction | Immovable Glass Sleeve |
| Material | Glass Shaft |
| pH Range | 0 to 14 |
| Type | Combination Electrode; Integrated Temperature Probe; Intelligent Sensor Management (ISM) Technology |
| For Use With (Application) | General laboratory applications |
| For Use With (Equipment) | Multi-Pin to BNC/RCA (30281896) |
| Internal Reference | ARGENTHAL™with Ag+-trap |
| Membrane | HA Glass |
| Model | InLab™ Max Pro-ISM |
| Show More |