Learn More
Fisherbrand™ Ink Chart Recorders for Isotemp Ultra-Low Temperature Chest Freezers
Description
- Seven-day recorder
- Functions to -150°C
- Factory installed
Specifications
Specifications
| Description | Factory Installed |
| Type | Chart Recorder |
| Temperature Range | -150°C |
| For Use With (Equipment) | CryoMed TSCM Series Freezers, Isotemp 13 and 20 cu. ft. Chest Ultra-Low Temperature Freezer |
| Static Holding Time | 7 Days |