Learn More
Com-Pac International™ Infecon Infectious Substance Shipper
Com-Pac International™ Infecon Infectious Substance Shipper has the best features of both an overpack shipper (INFECON 6000) and a UN Certified Category A shipper (INFECON 2000).
Supplier:  Com-Pac International™ INF6200
 View more versions of this product
Catalog No. 22-043-316
Only null left
Description
Description
  • Combination of both units and provides a shipping solution for any Category A or B specimen.
  • Vessel in its carton is to be placed in the overpack cooler, with refrigerated gel packs or dry ice.
  • Designed to ship Ebola, Enterovirus, Zika Virus, and Measles.
  • Included in the kit are specific instructions for shipment of Ebola and Enterovirus.
  • Can, however, be used to ship any Category A or B specimen.
  • Temperature protection for up to 76 hours
  • Holds approximately 22 pounds of Dry Ice
  • Expanded Polystyrene Insulated Tub
  • Sturdy Outer Box
  • Infectious Substance
  • Dry Ice Super Label
Specifications
Specifications
4/case
Cooler, Shipping Box, Stabilizer Sheet, Instruction Sheets, Package Return Label, Dry Ice Super Label, Infectious Substance Dry Ice Super Label, 1/2 Liter Pressure Vessel, 2 x 2 Biological Substance Label, 2 x 2 Infectious Substance Label, Shippers Declarations, 50 mL Capacity Absorbent Sheet 3 x 3, Perforated 6 Compartment Bubble Pouch
15.5 x 14 x 12.75 in.
Holds up to 6 standard blood tubes
Product Suggestions
Product Suggestions