Learn More
Fisherbrand™ Infecon™ 3000 Infectious Substance Shipper
Catalog No. 19037436
Change view
Click to view available options
Type:
Infectious Substance Shipper
Catalog No. Type
19037436 Infectious Substance Shipper
1 options
Catalog No. 19037436 Supplier Fisherbrand™ Supplier No. INF3000
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Combines U.N. certification requirements with new innovative safety features
  • Each shipper consists of a transparent pressure vessel able to withstand up to 95kPa, cushioning materials, outer box, labels, declarations and instructions
  • Transparent pressure vessel allows contents to be seen
  • Use the Infecon 3000 with the Speci-Freez™ Insulated Shipper (19-075-384C) for refrigerated or frozen specimens
  • Reusable when using the complete refurbishment kit (19-037-437)
Order Info
Dimensions: 6 x 6 x 8 in. (15.24 x 15.24 x 20.32cm) with 1L vessel
Specifications
Applications For infectious substances
Type Infectious Substance Shipper