Learn More
Eisco™ Inertia Demonstration Kit
Demonstrates inertia in a simple experiment that is suitable for middle school, high school, or even basic college physics.
Supplier:  Eisco™ PH0355IA
Catalog No. S27666
Only null left
Description
Description
  • Includes stand with a receptacle for a ball at the top
  • Pulling and releasing the trigger causes the square sheet to slide out from under the ball, dropping the ball in the receptacle by inertia
  • Suitable for middle school, high school, or early undergraduate physics classes
Specifications
Specifications
Kit, Inertia Demonstration
Physics Demontrations
18 mm Ball, Square Piece of Sheet Metal, Trigger of Sheet Metal, and Stand
Product Suggestions
Product Suggestions