Learn More
Supertek™ Inertia Balance Kit
Description
- Used for investigating the nature of mass and the distinction between mass and weight.
- Two metal trays 130 x 55 x 23 mm are joined together with two spring steel strips.
- One tray is drilled to accept three equal cylindrical masses
Specifications
Specifications
| Product Type | Archimedes Balance Set Demonstration |
| Dimensions (L x W x H) | 130 x 55 x 23 mm |