missing translation for 'onlineSavingsMsg'
Learn More

Indoxyl-beta-D-glucoside, 98%

Catalog No. AC458882500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC458882500 250 mg
AC458880010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC458882500 Supplier Thermo Scientific Chemicals Supplier No. 458882500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 487-60-5 | C14H17NO6 | 295.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 487-60-5
Molecular Formula C14H17NO6
Molecular Weight (g/mol) 295.29
MDL Number MFCD00047169
InChI Key XVARCVCWNFACQC-RKQHYHRCSA-N
Synonym indican, 3-indolyl-b-d-glucopyranoside, uroxanthin, 3-indoxyl-beta-d-glucopyranoside, indoxyl, a-d-glucoside, unii-n187wk1y1j, indikan, indican, plant, indican glucoside, 3-indolyl b-d-glucopyranoside
PubChem CID 441564
ChEBI CHEBI:16700
IUPAC Name (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-(1H-indol-3-yloxy)oxane-3,4,5-triol
SMILES OC[C@H]1O[C@@H](OC2=CNC3=CC=CC=C23)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

Melting Point 57.0°C to 58.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 250 mg
Solubility Information Solubility in water: freely soluble. Other solubilities: soluble in methanol, alcohol, acetone, slightly soluble in ether, benzene, chloroform,, ethyl acetate, carbon disulfide
Formula Weight 295.29
Percent Purity 98%
Chemical Name or Material Indoxyl-β-D-glucoside

RUO – Research Use Only