missing translation for 'onlineSavingsMsg'
Learn More

Indoxyl-beta-D-glucoside, 98%

CAS: 487-60-5 | C14H17NO6 | 295.29 g/mol

Supplier:  Thermo Scientific Chemicals 458880010

 View more versions of this product

Catalog No. AC458880010


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Indoxyl-β-D-glucoside
57.0°C to 58.0°C
97.5% min. (HPLC)
C14H17NO6
1 g
Solubility in water: freely soluble. Other solubilities: soluble in methanol, alcohol, acetone, slightly soluble in ether, benzene, chloroform,, ethyl acetate, carbon disulfide
OC[C@H]1O[C@@H](OC2=CNC3=CC=CC=C23)[C@H](O)[C@@H](O)[C@@H]1O
295.29
CHEBI:16700
98%
487-60-5
Authentic
Glass Bottle
MFCD00047169
indican, 3-indolyl b-d-glucopyranoside, uroxanthin, uroxanthin, indoxyl, indoxyl, unii-n187wk1y1j, unii-n187wk1y1j, indikan, indican glucoside, indican glucoside
XVARCVCWNFACQC-RKQHYHRCSA-N
(2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-(1H-indol-3-yloxy)oxane-3,4,5-triol
441564
295.29

Chemical Identifiers

487-60-5
295.29
XVARCVCWNFACQC-RKQHYHRCSA-N
441564
(2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-(1H-indol-3-yloxy)oxane-3,4,5-triol
C14H17NO6
MFCD00047169
indican, 3-indolyl b-d-glucopyranoside, uroxanthin, uroxanthin, indoxyl, indoxyl, unii-n187wk1y1j, unii-n187wk1y1j, indikan, indican glucoside, indican glucoside
CHEBI:16700
OC[C@H]1O[C@@H](OC2=CNC3=CC=CC=C23)[C@H](O)[C@@H](O)[C@@H]1O
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only