missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ Indoor Drum Storage Cabinet
GREENER_CHOICE

Catalog No. p-27280
Change view
Click to view available options
Dimensions (D x W x H) Interior:
30.34 x 30.75 x 60.5 in.
Width (Metric) Exterior:
86.4 cm
Weight (Metric):
177 kg
Certifications/Compliance:
FM, NFPA, OSHA
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (D x W x H) Interior Width (Metric) Exterior Weight (Metric) Certifications/Compliance
17151A 30.34 x 30.75 x 60.5 in. 86.4 cm 177 kg FM, NFPA, OSHA
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17151A Supplier Justrite™ Supplier No. 896200
Only null left

Justrite's Indoor Drum Storage Cabinet features a removable, adjustable, half-depth shelf with enough headspace to accommodate a drum with a pump or funnel. This means the cabinet can be used as a pumping or waste accumulation station , storage for Class III materials, or for stowing of spare accessories.

  • Double-wall 18-gauge steel with 1.5 in. (3.8cm) insulating air space
  • Three-point self-latching manual doors with double key set
  • Dual 2 in. (5cm) vents with flame arresters
  • Spill-catcher shelf adjustable on 2.25 in. (5.7cm) centers
  • Adjustable leveling feet
  • High-visibility warning label
  • 2 in. (5cm) leakproof sill to capture spills
  • Welded for superior strength
  • Powder-paint finish for maximum durability and chemical resistance

Order Info

Two capacities: hold either one or two 55-gal. (208L) drums.

Certifications

FM approved (except 17-150B).

Compliance

All meet NFPA and OSHA requirements.

Specifications

Dimensions (D x W x H) Interior 30.34 x 30.75 x 60.5 in.
Height (Metric) Exterior 165.1 cm
Weight (Metric) 177 kg
Width (Metric) Interior 78 cm
Height (Metric) Interior 153.7 cm
Width (Metric) Exterior 86.4 cm
No. of Doors 2
Depth (Metric) Interior 77.1 cm
No. of Shelves 1
Load Bearing Capacity 55 gal.
Depth (Metric) Exterior 86.4 cm
Product Type Vertical drum storage cabinet
Weight (English) 391 lb.
Door Style Manual-close
Certifications/Compliance FM, NFPA, OSHA
Height (English) Interior 60.5 in.
Material Steel (CR)
Depth (English) Interior 30.34 in.
Includes Half-depth shelf, installed pumps, drum support, level adjusting feet and hazardous warning label
Width (English) Interior 30.75 in.
Width (English) Exterior 34 in.
Height (English) Exterior 65 in.
For Use With (Application) Flammables
Color Yellow
Depth (English) Exterior 34 in.
Show More Show Less