missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Indomethacin

Catalog No. p-7077709
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Catalog No. Quantity
AC458030050 5 g
AC458030250 25 g
AC458031000 100 g
3 options

Catalog No. AC458030050

Supplier: Thermo Scientific Chemicals 458030050

Only null left

Indomethacin, CAS # 53-86-1, is an indole-based nonsteroidal anti-inflammatory (NSAID). | CAS: 53-86-1 | C19H16ClNO4 | 357.79 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Indomethacin is a synthetic NSAID of the indole family that inhibits cyclooxygenase and reduces prostaglandin and pyrogen release

• Indomethacin may suppress the multi-drug resistant transporter and thereby increase the efficacy of antineoplastics

• Indomethacin activates phosphatases that inhibit signaling pathways associated with cancer cell migration and proliferation

Applications

• Indomethacin may be used for the laboratory study of cancer cell proliferation, tumorigenesis and drug interactions

• Indomethacin may be used for laboratory study of the effect of inhibiting the multi-drug resistance transporter, or for downregulation of survival signaling in cells via survivin

Chemical Identifiers

CAS 53-86-1
Molecular Formula C19H16ClNO4
Molecular Weight (g/mol) 357.79
InChI Key CGIGDMFJXJATDK-UHFFFAOYSA-N
Synonym indomethacin, indometacin, indocin, indometacine, indomethacine, indocid, metindol, amuno, indomethazine, imbrilon
PubChem CID 3715
ChEBI CHEBI:49662
IUPAC Name 2-[1-(4-chlorobenzoyl)-5-methoxy-2-methylindol-3-yl]acetic acid
SMILES CC1=C(C2=C(N1C(=O)C3=CC=C(C=C3)Cl)C=CC(=C2)OC)CC(=O)O

Specifications

Melting Point 155.0°C
Loss on Drying 0.5% max. (100°C, 2 h, 5 mm Hg)
Infrared Spectrum Authentic
Quantity 5 g
Formula Weight 357.79
Percent Purity ≥97.5% (HPLC)
Chemical Name or Material Indomethacin
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal if swallowed.
May cause an allergic skin reaction.
May damage fertility.
May damage the unborn child.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Rinse mouth.
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
Obtain special instructions before use
EINECSNumber 200-186-5

RUO – Research Use Only