missing translation for 'onlineSavingsMsg'
Learn More

Indometacin Farnesil 98.0+%, TCI America™

Catalog No. I094325MG
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I094325MG 25 mg
I0943100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I094325MG Supplier TCI America Supplier No. I094325MG

Chemical Identifiers

CAS 85801-02-1
Molecular Formula C34H40ClNO4
Molecular Weight (g/mol) 562.147
MDL Number MFCD00870661
InChI Key CFIGYZZVJNJVDQ-LMJOQDENSA-N
PubChem CID 5282183
IUPAC Name [(2E,6E)-3,7,11-trimethyldodeca-2,6,10-trienyl] 2-[1-(4-chlorobenzoyl)-5-methoxy-2-methylindol-3-yl]acetate
SMILES CC1=C(C2=C(N1C(=O)C3=CC=C(C=C3)Cl)C=CC(=C2)OC)CC(=O)OCC=C(C)CCC=C(C)CCC=C(C)C

Specifications

Color Yellow
Quantity 25 mg
Formula Weight 562.15
Percent Purity ≥98.0% (HPLC)
Physical Form Liquid
Chemical Name or Material Indometacin Farnesil
RTECSNumber NL3522820
TSCA No