missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Indolo[3,2,1-jk]carbazole 98.0+%, TCI America™
$240.46 - $715.79
Chemical Identifiers
| CAS | 205-95-8 |
|---|---|
| Molecular Formula | C18H11N |
| Molecular Weight (g/mol) | 241.293 |
| InChI Key | SZLNOBJKCVERBJ-UHFFFAOYSA-N |
| PubChem CID | 15352087 |
| SMILES | C1=CC=C2C(=C1)C3=C4N2C5=CC=CC=C5C4=CC=C3 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
I1061200MG
|
TCI America
I1061200MG |
200 mg |
Each for $240.46
|
|
|||||
|
I10611G
|
TCI America
I10611G |
1 g |
Each for $715.79
|
|
|||||
Chemical Identifiers
| 205-95-8 | |
| 241.293 | |
| 15352087 |
| C18H11N | |
| SZLNOBJKCVERBJ-UHFFFAOYSA-N | |
| C1=CC=C2C(=C1)C3=C4N2C5=CC=CC=C5C4=CC=C3 |
Specifications
| 205-95-8 | |
| White-Yellow | |
| 200 mg | |
| C1=CC=C2C(=C1)C3=C4N2C5=CC=CC=C5C4=CC=C3 | |
| 15352087 | |
| ≥98.0% (GC) | |
| Indolo[3,2,1-jk]carbazole |
| 136°C | |
| C18H11N | |
| SZLNOBJKCVERBJ-UHFFFAOYSA-N | |
| 241.293 | |
| 241.29 | |
| Crystalline Powder |
Safety and Handling
TSCA : No