missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-Indoline-2-carboxylic acid, 95%

Catalog No. p-7028246
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2545006 5 g
AAB2545014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2545006 Supplier Thermo Scientific Chemicals Supplier No. B2545006
Only null left

CAS: 78348-24-0 | C9H8NO2 | 162.17 g/mol

(±)-Indoline-2-carboxylic acid is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(+/-)-Indoline-2-carboxylic acid is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 78348-24-0
Molecular Formula C9H8NO2
Molecular Weight (g/mol) 162.17
MDL Number MFCD00010635
InChI Key QNRXNRGSOJZINA-QMMMGPOBSA-M
Synonym indoline-2-carboxylic acid, +/--indoline-2-carboxylic acid, dl-indoline-2-carboxylic acid, 1h-indole-2-carboxylicacid, 2,3-dihydro, 2,3-dihydroindole-2-carboxylic acid, 1h-indole-2-carboxylic acid, 2,3-dihydro, 2-indolinecarboxylic acid, indoline-2-carboxylicacid, 1-indoline-2-carboxylic acid, pubchem3035
PubChem CID 86074
IUPAC Name 2,3-dihydro-1H-indole-2-carboxylic acid
SMILES [O-]C(=O)[C@@H]1CC2=CC=CC=C2N1

Specifications

Melting Point ∼168°C (decomposition)
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 163.18
Percent Purity 95%
Chemical Name or Material (+/-)-Indoline-2-carboxylic acid
Hazard Category H317-H361
Hazard Statement GHS H Statement
H361-H373-H317
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
EINECSNumber 278-899-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only