missing translation for 'onlineSavingsMsg'
Learn More

Indole-7-carboxaldehyde, 98%

Catalog No. p-7047064
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL17506MD 250 mg
AAL1750603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL17506MD Supplier Thermo Scientific Chemicals Supplier No. L17506MD

CAS: 1074-88-0 | C9H7NO | 145.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1074-88-0
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.16
MDL Number MFCD01318152
InChI Key XQVZDADGTFJAFM-UHFFFAOYSA-N
Synonym indole-7-carboxaldehyde, 7-formylindole, indole-7-carbaldehyde, 7-formyl-1h-indole, 1h-indole-7-carboxaldehyde, indole-7-carboxaldehyde 7-formylindole, 7-formyl indole, 7-indolcarboxaldehyde, pubchem7270, 7-indolecarboxaldehyde
PubChem CID 2734629
IUPAC Name 1H-indole-7-carbaldehyde
SMILES O=CC1=C2NC=CC2=CC=C1

Specifications

Melting Point 86°C to 89°C
Quantity 250 mg
Beilstein 1524960
Sensitivity Air Sensitive
Formula Weight 145.16
Percent Purity 98%
Chemical Name or Material Indole-7-carboxaldehyde
Hazard Category H302+H312-H315-H317-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only