missing translation for 'onlineSavingsMsg'
Learn More

Indole-6-boronic acid, 98%

Catalog No. p-7035594
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH53313MD 250 mg
AAH5331303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH53313MD Supplier Thermo Scientific Chemicals Supplier No. H53313MD

CAS: 147621-18-9 | C8H8BNO2 | 160.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 147621-18-9
Molecular Formula C8H8BNO2
Molecular Weight (g/mol) 160.97
MDL Number MFCD03095176
InChI Key ZVMHOIWRCCZGPZ-UHFFFAOYSA-N
Synonym indole-6-boronic acid, 6-indolylboronic acid, 1h-indol-6-yl boronic acid, 6-indoleboronic acid, indole-6-boronicacid, 1h-indol-6-yl-6-boronic acid, 6-borono-1h-indole, 6-indole boronic acid, 6-boronoindole, 1h-indole-6-boronic acid
PubChem CID 2763205
IUPAC Name 1H-indol-6-ylboronic acid
SMILES OB(O)C1=CC=C2C=CNC2=C1

Specifications

Melting Point 177°C to 181°C
Quantity 250 mg
Sensitivity Air Sensitive
Formula Weight 160.97
Percent Purity 98%
Chemical Name or Material Indole-6-boronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Store at -20°C; Store under Nitrogen

RUO – Research Use Only