missing translation for 'onlineSavingsMsg'
Learn More

Acide indole-5-carboxylique 98,0+%, TCI America™

Numéro de catalogue. I00295G
Modifier l'affichage
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
I00295G 5 g
I00291G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. I00295G Fournisseur TCI America Code fournisseur I00295G

Identifiants chimiques

CAS 1670-81-1
Molecular Formula C9H6NO2
Molecular Weight (g/mol) 160.15
MDL Number MFCD00005678
InChI Key IENZCGNHSIMFJE-UHFFFAOYSA-M
Synonym indole-5-carboxylic acid, 5-carboxyindole, 5-indolecarboxylic acid, indole-5-carboxylic aicd, pubchem1694, 5-carboxy-1h-indole, indole-5-carboxylicacid, indol-5-carboxylic acid, 5-indole carboxylic acid, indole 5-carboxylic acid
PubChem CID 74280
IUPAC Name 1H-indole-5-carboxylate
SMILES [O-]C(=O)C1=CC=C2NC=CC2=C1

Spécifications

Melting Point 212°C
Quantity 5 g
Formula Weight 161.16
Percent Purity ≥98.0% (GC,T)
Physical Form Poudre cristalline
Chemical Name or Material Indole-5-carboxylic Acid