missing translation for 'onlineSavingsMsg'
Learn More

Indole-5-carboxylic acid, 98%

Catalog No. p-148500
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC122210010 1 g Glass bottle
AC122210050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC122210010 Supplier Thermo Scientific Chemicals Supplier No. 122210010
Only null left

CAS: 1670-81-1 | C9H6NO2 | 160.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1670-81-1
Molecular Formula C9H6NO2
Molecular Weight (g/mol) 160.15
MDL Number MFCD00005678
InChI Key IENZCGNHSIMFJE-UHFFFAOYSA-M
Synonym indole-5-carboxylic acid, 5-carboxyindole, 5-indolecarboxylic acid, indole-5-carboxylic aicd, pubchem1694, 5-carboxy-1h-indole, indole-5-carboxylicacid, indol-5-carboxylic acid, 5-indole carboxylic acid, indole 5-carboxylic acid
PubChem CID 74280
IUPAC Name 1H-indole-5-carboxylic acid
SMILES [O-]C(=O)C1=CC=C2NC=CC2=C1

Specifications

Melting Point 207.0°C to 212.0°C
Color Beige to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 161.16
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Indole-5-carboxylic acid, 98%
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
Harmful in contact with skin.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 216-799-6

RUO – Research Use Only