missing translation for 'onlineSavingsMsg'
Learn More

Indole-5-carboxaldehyde, 98+%

Catalog No. AC391630250
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC391630250 25 g Glass bottle
AC391630010 1 g Glass bottle
AC391630050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC391630250 Supplier Thermo Scientific Chemicals Supplier No. 391630250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1196-69-6 | C9H7NO | 145.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1196-69-6
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.16
MDL Number MFCD02093664
InChI Key ADZUEEUKBYCSEY-UHFFFAOYSA-N
Synonym indole-5-carboxaldehyde, 5-formylindole, indole-5-carbaldehyde, 1h-indole-5-carboxaldehyde, 5-indolealdehyde, indole-5-aldehyde, 5-formyl-1h-indole, 5-indolecarboxaldehyde, indole-5-carboxaldehyde 5-formylindole, 5-formyl-indole
PubChem CID 589040
IUPAC Name 1H-indole-5-carbaldehyde
SMILES C1=CC2=C(C=CN2)C=C1C=O

Specifications

Melting Point 100°C to 103°C
Color Orange or Tan
Infrared Spectrum Authentic
Assay Percent Range 98% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Formula Weight 145.16
Percent Purity 98+%
Physical Form Powder
Chemical Name or Material Indole-5-carboxaldehyde
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only