missing translation for 'onlineSavingsMsg'
Learn More

Indole-5-carboxaldehyde, 98%

Catalog No. AAB2551706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2551706 5 g
AAB2551703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2551706 Supplier Thermo Scientific Chemicals Supplier No. B2551706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1196-69-6 | C9H7NO | 145.161 g/mol

Indole-5-carboxaldehyde is used as a reactant in preparation of curcumin derivatives as anti-proliferative & anti-inflammatory agents; and in synthesis of para-para stilbenophanes by McMurry coupling. It is also used as a reactant in stereoselective synthesis of dibenzylideneacetone derivatives as β-amyloid imaging probes and in structure-based drug design of aurora kinase A inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Indole-5-carboxaldehyde is used as a reactant in preparation of curcumin derivatives as anti-proliferative & anti-inflammatory agents; and in synthesis of para-para stilbenophanes by McMurry coupling. It is also used as a reactant in stereoselective synthesis of dibenzylideneacetone derivatives as β-amyloid imaging probes and in structure-based drug design of aurora kinase A inhibitors.

Solubility
Insoluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 1196-69-6
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.161
MDL Number MFCD02093664
InChI Key ADZUEEUKBYCSEY-UHFFFAOYSA-N
Synonym indole-5-carboxaldehyde, 5-formylindole, indole-5-carbaldehyde, 1h-indole-5-carboxaldehyde, 5-indolealdehyde, indole-5-aldehyde, 5-formyl-1h-indole, 5-indolecarboxaldehyde, indole-5-carboxaldehyde 5-formylindole, 5-formyl-indole
PubChem CID 589040
IUPAC Name 1H-indole-5-carbaldehyde
SMILES C1=CC2=C(C=CN2)C=C1C=O

Specifications

Melting Point 96°C to 100°C
Quantity 5 g
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 145.16
Percent Purity 98%
Chemical Name or Material Indole-5-carboxaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only