missing translation for 'onlineSavingsMsg'
Learn More

Indole-4-carboxaldehyde 98.0+%, TCI America™

Catalog No. I07125G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I07125G 5 g
I07121G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I07125G Supplier TCI America Supplier No. I07125G
Only null left

Chemical Identifiers

CAS 1074-86-8
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.16
MDL Number MFCD01632221
InChI Key JFDDFGLNZWNJTK-UHFFFAOYSA-N
Synonym indole-4-carboxaldehyde, 4-formylindole, 1h-indole-4-carboxaldehyde, indole-4-carbaldehyde, 4-indole-carboxaldehyde, 4-indolecarbaldehyde, 4-formyl-indole, 4-formyl-1h-indole, 1h-indol-4-carbaldehyde, indole-4-aldehyde
PubChem CID 333703
IUPAC Name 1H-indole-4-carbaldehyde
SMILES O=CC1=C2C=CNC2=CC=C1

Specifications

Melting Point 144°C
Quantity 5 g
Formula Weight 145.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Indole-4-carboxaldehyde