missing translation for 'onlineSavingsMsg'
Learn More

Indole-3-carboxaldehyde 98.0+%, TCI America™

$69.31 - $179.05

Chemical Identifiers

CAS 487-89-8
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.161
MDL Number MFCD00005622
InChI Key OLNJUISKUQQNIM-UHFFFAOYSA-N
Synonym indole-3-carboxaldehyde, 3-formylindole, 1h-indole-3-carboxaldehyde, indole-3-aldehyde, indole-3-carbaldehyde, indole-3-carboxyaldehyde, 3-indolecarboxaldehyde, beta-indolylaldehyde, 3-indolecarbaldehyde, indol-3-carboxaldehyde
PubChem CID 10256
ChEBI CHEBI:28238
IUPAC Name 1H-indole-3-carbaldehyde
SMILES C1=CC=C2C(=C1)C(=CN2)C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I00275G
View Documents
TCI America
I00275G
5 g
Each for $69.31
Only null left
 
I002725G
View Documents
TCI America
I002725G
25 g
Each for $179.05
Only null left
 
Description

Description

Specifications

Chemical Identifiers

487-89-8
145.161
OLNJUISKUQQNIM-UHFFFAOYSA-N
10256
1H-indole-3-carbaldehyde
C9H7NO
MFCD00005622
indole-3-carboxaldehyde, 3-formylindole, 1h-indole-3-carboxaldehyde, indole-3-aldehyde, indole-3-carbaldehyde, indole-3-carboxyaldehyde, 3-indolecarboxaldehyde, beta-indolylaldehyde, 3-indolecarbaldehyde, indol-3-carboxaldehyde
CHEBI:28238
C1=CC=C2C(=C1)C(=CN2)C=O

Specifications

487-89-8
C9H7NO
5 g
OLNJUISKUQQNIM-UHFFFAOYSA-N
1H-indole-3-carbaldehyde
10256
145.16
Crystalline Powder
195°C
MFCD00005622
indole-3-carboxaldehyde, 3-formylindole, 1h-indole-3-carboxaldehyde, indole-3-aldehyde, indole-3-carbaldehyde, indole-3-carboxyaldehyde, 3-indolecarboxaldehyde, beta-indolylaldehyde, 3-indolecarbaldehyde, indol-3-carboxaldehyde
C1=CC=C2C(=C1)C(=CN2)C=O
145.161
CHEBI:28238
≥98.0% (GC)
Indole-3-carboxaldehyde
Videos
Safety and Handling

Safety and Handling

RTECSNumber : NL5993600

TSCA : Yes

SDS
Documents

Documents