missing translation for 'onlineSavingsMsg'
Learn More

Indole-3-carboxaldehyde 98.0+%, TCI America™

Catalog No. I00275G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I00275G 5 g
I002725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I00275G Supplier TCI America Supplier No. I00275G

Chemical Identifiers

CAS 487-89-8
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.161
MDL Number MFCD00005622
InChI Key OLNJUISKUQQNIM-UHFFFAOYSA-N
Synonym indole-3-carboxaldehyde, 3-formylindole, 1h-indole-3-carboxaldehyde, indole-3-aldehyde, indole-3-carbaldehyde, indole-3-carboxyaldehyde, 3-indolecarboxaldehyde, beta-indolylaldehyde, 3-indolecarbaldehyde, indol-3-carboxaldehyde
PubChem CID 10256
ChEBI CHEBI:28238
IUPAC Name 1H-indole-3-carbaldehyde
SMILES C1=CC=C2C(=C1)C(=CN2)C=O

Specifications

Melting Point 195°C
Quantity 5 g
Formula Weight 145.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Indole-3-carboxaldehyde
RTECSNumber NL5993600
TSCA Yes