missing translation for 'onlineSavingsMsg'
Learn More

Indole-3-carboxaldehyde, 99%

Catalog No. AAA1533014
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1533014 25 g
AAA1533022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1533014 Supplier Thermo Scientific Chemicals Supplier No. A1533014

CAS: 487-89-8 | C9H7NO | 145.161 g/mol

Indole-3-carboxaldehyde is a biochemical used to prepare analogs of the indole phytoalexin cyclobrassinin with NR1R2 group. It was also used as the starting material for the synthesis of higher order indoles including isoindolo[2,1-a]indoles, aplysinopsins, and 4-substituted-tetrahydrobenz[cd]indoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Indole-3-carboxaldehyde is a biochemical used to prepare analogs of the indole phytoalexin cyclobrassinin with NR1R2 group. It was also used as the starting material for the synthesis of higher order indoles including isoindolo[2,1-a]indoles, aplysinopsins, and 4-substituted-tetrahydrobenz[cd]indoles.

Solubility
Insoluble in water.

Notes
Air Sensitive. Avoid contact with air/oxygen. Incompatible with reducing agents.

Chemical Identifiers

CAS 487-89-8
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.161
MDL Number MFCD00005622
InChI Key OLNJUISKUQQNIM-UHFFFAOYSA-N
Synonym indole-3-carboxaldehyde, 3-formylindole, 1h-indole-3-carboxaldehyde, indole-3-aldehyde, indole-3-carbaldehyde, indole-3-carboxyaldehyde, 3-indolecarboxaldehyde, beta-indolylaldehyde, 3-indolecarbaldehyde, indol-3-carboxaldehyde
PubChem CID 10256
ChEBI CHEBI:28238
IUPAC Name 1H-indole-3-carbaldehyde
SMILES C1=CC=C2C(=C1)C(=CN2)C=O

Specifications

Melting Point 195°C to 198°C
Flash Point 240°C (464°F)
Quantity 25 g
Beilstein 114117
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 145.16
Percent Purity 99%
Chemical Name or Material Indole-3-carboxaldehyde
EINECSNumber 207-665-8
RTECSNumber NL5993600
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only