missing translation for 'onlineSavingsMsg'
Learn More

Indole-3-carboxaldehyde, 97%

Catalog No. AC156640050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC156640050 5 g Glass bottle
AC156640250 25 g Plastic bottle
AC156641000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC156640050 Supplier Thermo Scientific Chemicals Supplier No. 156640050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 487-89-8 | C9H7NO | 145.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 487-89-8
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.16
MDL Number MFCD00005622
InChI Key OLNJUISKUQQNIM-UHFFFAOYSA-N
Synonym indole-3-carboxaldehyde, 3-formylindole, 1h-indole-3-carboxaldehyde, indole-3-aldehyde, indole-3-carbaldehyde, indole-3-carboxyaldehyde, 3-indolecarboxaldehyde, beta-indolylaldehyde, 3-indolecarbaldehyde, indol-3-carboxaldehyde
PubChem CID 10256
ChEBI CHEBI:28238
IUPAC Name 1H-indole-3-carbaldehyde
SMILES C1=CC=C2C(=C1)C(=CN2)C=O

Specifications

Melting Point 193°C to 199°C
Flash Point 240°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 21, 313
Solubility Information Solubility in water: insoluble
Formula Weight 145.16
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Indole-3-carboxaldehyde, 97%
Show More Show Less
Hazard Category AIR SENSITIVE
EINECSNumber 207-665-8
RTECSNumber NL5993600
TSCA TSCA

RUO – Research Use Only