missing translation for 'onlineSavingsMsg'
Learn More

Indole-3-Butyric Acid Potassium Salt, MP Biomedicals™

Catalog No. ICN10204301
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10204301 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10204301 Supplier MP Biomedicals Inc Supplier No. 0210204301
Only null left

Synonym: K-IBA, Potassium 4-(1H-Indol-3-yl)butanoate, 1H-Indole-3-Butanoic Acid Potassium Salt

Water-soluble form of IBA, an auxin-family plant growth hormone.

  • Promotes root organogenesis
  • Induces callus formation
  • Forms adventitious roots
  • Aids in regulation of gravitropism and phototropism

Chemical Identifiers

Molecular Formula C12H12KNO2
Molecular Weight (g/mol) 241.331
MDL Number MFCD00058442
InChI Key KTWDHJYSJOSTSJ-UHFFFAOYSA-M
Synonym potassium 4-1h-indol-3-yl butanoate, indole-3-butyric acid potassium salt, indole-3-butyric acid potassium, 4-indol-3-ylbutanoic acid, potassium salt, potassium indolebutyrate, pubchem22140, cambridge id 5119461, potassium ion indole-3 butyrate, indole-3-butyric acid-potassium salt, 3-indole butyric acid, potassium salt
PubChem CID 23664348
IUPAC Name potassium;4-(1H-indol-3-yl)butanoate
SMILES C1=CC=C2C(=C1)C(=CN2)CCCC(=O)[O-].[K+]

Specifications

CAS 60096-23-3
Quantity 1 g
Physical Form Powder

Unless specified otherwise, MP Biomedical™'s products are for research or further manufacturing use only, not for direct human use.