missing translation for 'onlineSavingsMsg'
Learn More

Indole-3-butyric acid, 98%

Catalog No. AAA1526022
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1526022 100 g
AAA1526006 5 g
AAA1526014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1526022 Supplier Thermo Scientific Chemicals Supplier No. A1526022
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 133-32-4 | C12H13NO2 | 203.241 g/mol

Indole-3-butyric acid is auxin-family plant hormone (phytohormone). IBA is thought to be a precursor of indole-3-acetic acid (IAA) the most abundant and the basic auxin natively occurring and functioning in plants. IAA generates the majority of auxin effects in intact plants, and is the most potent native auxin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Indole-3-butyric acid is auxin-family plant hormone (phytohormone). IBA is thought to be a precursor of indole-3-acetic acid (IAA) the most abundant and the basic auxin natively occurring and functioning in plants. IAA generates the majority of auxin effects in intact plants, and is the most potent native auxin.

Solubility
Soluble in water(0.25g/L).

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 133-32-4
Molecular Formula C12H13NO2
Molecular Weight (g/mol) 203.241
MDL Number MFCD00005664
InChI Key JTEDVYBZBROSJT-UHFFFAOYSA-N
Synonym indole-3-butyric acid, 3-indolebutyric acid, 4-1h-indol-3-yl butanoic acid, indolebutyric acid, hormodin, 1h-indole-3-butanoic acid, seradix, indole-3-butanoic acid, jiffy grow, 4-indol-3-yl butyric acid
PubChem CID 8617
ChEBI CHEBI:33070
IUPAC Name 4-(1H-indol-3-yl)butanoic acid
SMILES C1=CC=C2C(=C1)C(=CN2)CCCC(=O)O

Specifications

Melting Point 121°C to 125°C
Odor Odorless
Assay Percent Range 98%
Quantity 100 g
UN Number UN2811
Beilstein 171120
Merck Index 14,4965
Solubility Information Soluble in water(0.25g/L).
Formula Weight 203.24
Percent Purity 98%
Chemical Name or Material Indole-3-butyric acid
Show More Show Less
Hazard Category H301-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 205-101-5
RTECSNumber NL5250000
TSCA Yes
storageNote1 Air sensitive
Recommended Storage Ambient temperatures

RUO – Research Use Only