missing translation for 'onlineSavingsMsg'
Learn More

Indole-3-butyric Acid, 97%, Spectrum™ Chemical

Catalog No. p-7165760
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611029 5 g Amber Glass Bottle
18611028 25 g Amber Glass Bottle
18611027 100 g Amber Glass Bottle
18611026 1 kg Poly Pail
18611030 5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18611029 Supplier Spectrum Chemical Mfg Cor Supplier No. IN1275GM
Only null left

Chemical Identifiers

CAS 133-32-4

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 5 g
Percent Purity 0.97
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.