missing translation for 'onlineSavingsMsg'
Learn More

Indole-3-butyric Acid, 97%, Spectrum™ Chemical

Catalog No. 18611030
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
5 kg
Packaging:
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611030 5 kg Poly Pail
18611029 5 g Poly Pail
18611028 25 g Poly Pail
18611027 100 g Poly Pail
18611026 1 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18611030 Supplier Spectrum Chemical Mfg Cor Supplier No. IN1275KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 133-32-4

Specifications

CAS Max % 1
Packaging Poly Pail
Quantity 5 kg
Molecular Weight (g/mol) 203.23
Percent Purity 0.97
Grade Ungraded
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Indole-3-butyric acid

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.