missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Indole-2-carboxylic Acid 98.0+%, TCI America™
$33.56 - $68.97
Chemical Identifiers
| CAS | 1477-50-5 |
|---|---|
| Molecular Formula | C9H7NO2 |
| Molecular Weight (g/mol) | 161.16 |
| MDL Number | MFCD00005611 |
| InChI Key | HCUARRIEZVDMPT-UHFFFAOYSA-N |
| Synonym | indole-2-carboxylic acid, 2-carboxyindole, 2-indolecarboxylic acid, indol-2-carboxylic acid, 1h-indolecarboxylic acid, indole-2-carboxylate, 2-indolylformic acid, 2-indole carboxylic acid, 1h-indol-2-carbons, carboxyindole |
| PubChem CID | 72899 |
| IUPAC Name | 1H-indole-2-carboxylic acid |
| SMILES | OC(=O)C1=CC2=CC=CC=C2N1 |
Chemical Identifiers
| 1477-50-5 | |
| 161.16 | |
| HCUARRIEZVDMPT-UHFFFAOYSA-N | |
| 72899 | |
| OC(=O)C1=CC2=CC=CC=C2N1 |
| C9H7NO2 | |
| MFCD00005611 | |
| indole-2-carboxylic acid, 2-carboxyindole, 2-indolecarboxylic acid, indol-2-carboxylic acid, 1h-indolecarboxylic acid, indole-2-carboxylate, 2-indolylformic acid, 2-indole carboxylic acid, 1h-indol-2-carbons, carboxyindole | |
| 1H-indole-2-carboxylic acid |
Specifications
| 1477-50-5 | |
| C9H7NO2 | |
| 5 g | |
| HCUARRIEZVDMPT-UHFFFAOYSA-N | |
| 1H-indole-2-carboxylic acid | |
| 72899 | |
| ≥98.0% (GC,T) | |
| Indole-2-carboxylic Acid |
| 205°C | |
| MFCD00005611 | |
| indole-2-carboxylic acid, 2-carboxyindole, 2-indolecarboxylic acid, indol-2-carboxylic acid, 1h-indolecarboxylic acid, indole-2-carboxylate, 2-indolylformic acid, 2-indole carboxylic acid, 1h-indol-2-carbons, carboxyindole | |
| OC(=O)C1=CC2=CC=CC=C2N1 | |
| 161.16 | |
| 161.16 | |
| Crystalline Powder |
Safety and Handling
TSCA : No