missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acide indole-2-carboxylique 98,0+%, TCI America™
Spécifications
Spécifications
| Melting Point | 205°C |
| Quantity | 5 g |
| Formula Weight | 161.16 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Poudre cristalline |
| Chemical Name or Material | Indole-2-carboxylic Acid |
Sécurité et manipulation
| TSCA | No |