missing translation for 'onlineSavingsMsg'
Learn More

Indole-2-carboxylic acid, 99%

CAS: 1477-50-5 | C9H7NO2 | 161.16 g/mol

$114.84 - $352.13

Chemical Identifiers

CAS 1477-50-5
Molecular Formula C9H7NO2
Molecular Weight (g/mol) 161.16
MDL Number MFCD00005611
InChI Key HCUARRIEZVDMPT-UHFFFAOYSA-N
Synonym indole-2-carboxylic acid, 2-carboxyindole, 2-indolecarboxylic acid, indol-2-carboxylic acid, 1h-indolecarboxylic acid, indole-2-carboxylate, 2-indolylformic acid, 2-indole carboxylic acid, 1h-indol-2-carbons, carboxyindole
PubChem CID 72899
IUPAC Name 1H-indole-2-carboxylic acid
SMILES OC(=O)C1=CC2=CC=CC=C2N1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC122200100
SDS View Documents
Thermo Scientific Chemicals
122200100
10 g Glass bottle
Each for $114.84
Only null left
 
AC122200500
SDS View Documents
Thermo Scientific Chemicals
122200500
50 g Glass bottle
Each for $352.13
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1477-50-5
161.16
HCUARRIEZVDMPT-UHFFFAOYSA-N
72899
OC(=O)C1=CC2=CC=CC=C2N1
C9H7NO2
MFCD00005611
indole-2-carboxylic acid, 2-carboxyindole, 2-indolecarboxylic acid, indol-2-carboxylic acid, 1h-indolecarboxylic acid, indole-2-carboxylate, 2-indolylformic acid, 2-indole carboxylic acid, 1h-indol-2-carbons, carboxyindole
1H-indole-2-carboxylic acid

Specifications

1477-50-5
Brown to White
98.5% min. (HPLC)
C9H7NO2
10 g
indole-2-carboxylic acid, 2-carboxyindole, 2-indolecarboxylic acid, indol-2-carboxylic acid, 1h-indolecarboxylic acid, indole-2-carboxylate, 2-indolylformic acid, 2-indole carboxylic acid, 1h-indol-2-carbons, carboxyindole
HCUARRIEZVDMPT-UHFFFAOYSA-N
1H-indole-2-carboxylic acid
72899
99%
Indole-2-carboxylic acid, 99%
205.0°C to 209.0°C
Authentic
Glass bottle
MFCD00005611
22, 61
Solubility in water: insoluble. Other solubilities: soluble in ethanol,ether
OC(=O)C1=CC2=CC=CC=C2N1
161.16
161.16
Crystalline Powder or Crystals
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 216-030-4

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only