missing translation for 'onlineSavingsMsg'
Learn More

Indole-2-carboxaldehyde 97.0+%, TCI America™

Catalog No. p-7131631
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I08521G 1 g
I08525G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I08521G Supplier TCI America Supplier No. I08521G
Only null left

Chemical Identifiers

CAS 19005-93-7
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.16
MDL Number MFCD03001425
InChI Key SBNOTUDDIXOFSN-UHFFFAOYSA-N
Synonym indole-2-carboxaldehyde, 1h-indole-2-carboxaldehyde, indole-2-carbaldehyde, 2-formylindole, 2-formyl-1h-indole, 1h-indolecarboxaldehyde, unii-r5n9cd7dlm, r5n9cd7dlm, methanol, 2h-indol-2-ylidene-, 1e-9ci, indolaldehyd
PubChem CID 96389
IUPAC Name 1H-indole-2-carbaldehyde
SMILES O=CC1=CC2=CC=CC=C2N1

Specifications

Melting Point 139°C
Quantity 1 g
Formula Weight 145.16
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Indole-2-carboxaldehyde