missing translation for 'onlineSavingsMsg'
Learn More

Indole-2-carboxaldehyde, 97%

Catalog No. p-7036360
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH54638MD 250 mg
AAH5463803 1 g
AAH5463806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH54638MD Supplier Thermo Scientific Chemicals Supplier No. H54638MD
Only null left

CAS: 19005-93-7 | C9H7NO | 145.16 g/mol

Indole-2-carboxaldehyde is a useful starter in indole and natural product chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Indole-2-carboxaldehyde is a useful starter in indole and natural product chemistry.

Solubility
Soluble in Methanol.

Notes
Air sensitive. Strong oxidizers and strong reducing agents are incompatible. Store at room temperature.

Chemical Identifiers

CAS 19005-93-7
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.16
MDL Number MFCD03001425
InChI Key SBNOTUDDIXOFSN-UHFFFAOYSA-N
Synonym indole-2-carboxaldehyde, 1h-indole-2-carboxaldehyde, indole-2-carbaldehyde, 2-formylindole, 2-formyl-1h-indole, 1h-indolecarboxaldehyde, unii-r5n9cd7dlm, r5n9cd7dlm, methanol, 2h-indol-2-ylidene-, 1e-9ci, indolaldehyd
PubChem CID 96389
IUPAC Name 1H-indole-2-carbaldehyde
SMILES O=CC1=CC2=CC=CC=C2N1

Specifications

Melting Point 138°C to 142°C
Quantity 250 mg
Sensitivity Air Sensitive
Solubility Information Soluble in Methanol.
Formula Weight 145.16
Percent Purity 97%
Chemical Name or Material Indole-2-carboxaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only