missing translation for 'onlineSavingsMsg'
Learn More

Indole-2-carbonitrile, 97%

Catalog No. p-7057101
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6645103 1 g
AAH6645106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6645103 Supplier Thermo Scientific Chemicals Supplier No. H6645103
Only null left

CAS: 36193-65-4 | C9H6N2 | 142.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 36193-65-4
Molecular Formula C9H6N2
Molecular Weight (g/mol) 142.16
MDL Number MFCD00187566
InChI Key CBTITARLOCZPDU-UHFFFAOYSA-N
Synonym 2-cyanoindole, cyanoindole, 2-indolecarbonitrile, 2-cyano-1h-indole, indole-2-carbonitrile, racemic indole nitrile
PubChem CID 3787599
IUPAC Name 1H-indole-2-carbonitrile
SMILES N#CC1=CC2=CC=CC=C2N1

Specifications

Quantity 1 g
UN Number UN3439
Beilstein 116664
Formula Weight 142.16
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Indole-2-carbonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only