missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Indoine Blue, Thermo Scientific™
Chemical Identifiers
| CAS | 4569-88-4 |
|---|---|
| Molecular Formula | C30H24ClN5O |
| Molecular Weight (g/mol) | 506.01 |
| MDL Number | MFCD00011924 |
| InChI Key | SCMDRBZEIUMBBQ-UHFFFAOYSA-N |
| PubChem CID | 9575841 |
| IUPAC Name | 3-amino-2,8-dimethyl-7-{2-[(1Z)-2-oxo-1,2-dihydronaphthalen-1-ylidene]hydrazin-1-yl}-5-phenyl-5λâµ-phenazin-5-ylium chloride |
| SMILES | [Cl-].CC1=CC2=NC3=CC(C)=C(N)C=C3[N+](C3=CC=CC=C3)=C2C=C1N\N=C1/C(=O)C=CC2=CC=CC=C12 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC365330010
|
Thermo Scientific Chemicals
365330010 |
1g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC365330050
|
Thermo Scientific Chemicals
365330050 |
5g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 4569-88-4 | |
| 506.01 | |
| SCMDRBZEIUMBBQ-UHFFFAOYSA-N | |
| 3-amino-2,8-dimethyl-7-{2-[(1Z)-2-oxo-1,2-dihydronaphthalen-1-ylidene]hydrazin-1-yl}-5-phenyl-5λâµ-phenazin-5-ylium chloride |
| C30H24ClN5O | |
| MFCD00011924 | |
| 9575841 | |
| [Cl-].CC1=CC2=NC3=CC(C)=C(N)C=C3[N+](C3=CC=CC=C3)=C2C=C1N\N=C1/C(=O)C=CC2=CC=CC=C12 |
Specifications
| 4569-88-4 | |
| MFCD00011924 | |
| Authentic | |
| 3-amino-2,8-dimethyl-7-{2-[(1Z)-2-oxo-1,2-dihydronaphthalen-1-ylidene]hydrazin-1-yl}-5-phenyl-5λâµ-phenazin-5-ylium chloride | |
| 9575841 | |
| 10% max. (110°C, 1 h) | |
| 1g |
| C30H24ClN5O | |
| SCMDRBZEIUMBBQ-UHFFFAOYSA-N | |
| [Cl-].CC1=CC2=NC3=CC(C)=C(N)C=C3[N+](C3=CC=CC=C3)=C2C=C1N\N=C1/C(=O)C=CC2=CC=CC=C12 | |
| 506.01 | |
| 595 to 601nm (0.001% in methanol) | |
| 506.01 | |
| Indoine Blue |
RUO â Research Use Only