missing translation for 'onlineSavingsMsg'
Learn More

Indoine Blue, Thermo Scientific™

Catalog No. AC365330010
Change view
Click to view available options
Quantity:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC365330010 1g
AC365330050 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC365330010 Supplier Thermo Scientific Chemicals Supplier No. 365330010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4569-88-4
Molecular Formula C30H24ClN5O
Molecular Weight (g/mol) 506.01
MDL Number MFCD00011924
InChI Key SCMDRBZEIUMBBQ-UHFFFAOYSA-N
PubChem CID 9575841
IUPAC Name 3-amino-2,8-dimethyl-7-{2-[(1Z)-2-oxo-1,2-dihydronaphthalen-1-ylidene]hydrazin-1-yl}-5-phenyl-5λ⁵-phenazin-5-ylium chloride
SMILES [Cl-].CC1=CC2=NC3=CC(C)=C(N)C=C3[N+](C3=CC=CC=C3)=C2C=C1N\N=C1/C(=O)C=CC2=CC=CC=C12

Specifications

Infrared Spectrum Authentic
Lambda Max 595 to 601nm (0.001% in methanol)
Formula Weight 506.01
Loss on Drying 10% max. (110°C, 1 h)
Quantity 1g
Chemical Name or Material Indoine Blue

RUO – Research Use Only