missing translation for 'onlineSavingsMsg'
Learn More

Indocyanine Green, TCI America™

Catalog No. p-7131467
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0535100MG 100 mg
I05351G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I0535100MG Supplier TCI America Supplier No. I0535100MG

Chemical Identifiers

CAS 3599-32-4
Molecular Formula C43H47N2NaO6S2
Molecular Weight (g/mol) 774.967
MDL Number MFCD00013078
InChI Key MOFVSTNWEDAEEK-UHFFFAOYSA-M
Synonym indocyanine green
PubChem CID 132274068
ChEBI CHEBI:31696
IUPAC Name sodium;4-[(2E)-2-[(2E,4E,6Z)-7-[1,1-dimethyl-3-(4-sulfonatobutyl)benzo[e]indol-3-ium-2-yl]hepta-2,4,6-trienylidene]-1,1-dimethylbenzo[e]indol-3-yl]butane-1-sulfonate
SMILES CC1(C(=[N+](C2=C1C3=CC=CC=C3C=C2)CCCCS(=O)(=O)[O-])C=CC=CC=CC=C4C(C5=C(N4CCCCS(=O)(=O)[O-])C=CC6=CC=CC=C65)(C)C)C.[Na+]

Specifications

Quantity 100 mg
Formula Weight 774.97
Physical Form Crystalline Powder
Chemical Name or Material Indocyanine Green
RTECSNumber DE3150000
TSCA Yes