missing translation for 'onlineSavingsMsg'
Learn More

Indium oxide, tin doped, polymeric precursor, Oxide ≈20 wt%

Catalog No. AA4222914
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4222914 25 g
AA4222909 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4222914 Supplier Thermo Scientific Chemicals Supplier No. 04222914

Preparation of transparent electrical conducting electrodes | CAS: 1312-43-2 | In2O3 | 277.63 g/mol

It is used in the preparation of transparent electrical conducting electrodes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1312-43-2
Molecular Formula In2O3
Molecular Weight (g/mol) 277.63
MDL Number MFCD00011060
InChI Key PJXISJQVUVHSOJ-UHFFFAOYSA-N
IUPAC Name diindium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[In+3].[In+3]

Specifications

Physical Form Viscous Liquid
Quantity 25 g
UN Number UN1993
Flash Point 13°C (55°F)
Solubility Information Soluble in hexane and toluene; Insoluble in alcohols
Concentration or Composition (by Analyte or Components) Oxide ≈20 wt%
Chemical Name or Material Indium oxide, Tin doped, polymeric precursor
Hazard Category H225-H319-H336-H372
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: II
TSCA Yes

RUO – Research Use Only