missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) sulfate hydrate, Puratronic™, 99.999% (metals basis)

Catalog No. p-7048505
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1071109 10 g
AA1071118 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1071109 Supplier Thermo Scientific Chemicals Supplier No. 01071109

CAS: 13464-82-9 | In2O12S3 | 517.80 g/mol

Indium(III) sulfate is used Chemical synthesis, hardening agent in gold electroplating baths. It is also used to prepare other indium containing substances such as copper indium selenide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13464-82-9
Molecular Formula In2O12S3
Molecular Weight (g/mol) 517.80
MDL Number MFCD00011059
InChI Key XGCKLPDYTQRDTR-UHFFFAOYSA-H
Synonym indium sulfate, indisulfat, indium sulphate, indium iii sulfate, diindium tris sulphate, indium trisulfate, diindium trisulfate, indium sequisulfate, sulfuric acid, indium salt, indisulfat german
PubChem CID 26044
IUPAC Name indium(3+);trisulfate
SMILES [In+3].[In+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Melting Point 250°C (decomposition)
Physical Form Lump/Powder
Quantity 10 g
Solubility Information Soluble in water and ethanol. Insoluble in basic salts.
Formula Weight 517.83 (Anhydrous)
Percent Purity 99.999%
Grade Puratronic
Assay (metals basis)
Chemical Name or Material Indium(III) sulfate hydrate
EINECSNumber 236-689-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only