missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) sulfate, anhydrous, 99.99% (metals basis)

Catalog No. AA4010214
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4010214 25 g
AA4010222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4010214 Supplier Thermo Scientific Chemicals Supplier No. 04010214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 13464-82-9 | In2O12S3 | 517.80 g/mol

Indium(III) sulfate, anhydrous is used in the preparation of indium or indium containing substances like copper indium selenide. It is employed to electroplate indium metal, as a hardening agent in gold electroplating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13464-82-9
Molecular Formula In2O12S3
Molecular Weight (g/mol) 517.80
MDL Number MFCD00011059
InChI Key XGCKLPDYTQRDTR-UHFFFAOYSA-H
Synonym indium sulfate, indisulfat, indium sulphate, indium iii sulfate, diindium tris sulphate, indium trisulfate, diindium trisulfate, indium sequisulfate, sulfuric acid, indium salt, indisulfat german
PubChem CID 26044
SMILES [In+3].[In+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Color Gray
Physical Form Solid
Quantity 25 g
Solubility Information Soluble in water.
IUPAC Name indium(3+);trisulfate
Formula Weight 517.83
Percent Purity 99.99%
Odor Odorless
Sensitivity Hygroscopic
Density 3.438 g/mL
Assay (metals basis)
Chemical Name or Material Indium(III) sulfate, Anhydrous
Show More Show Less
EINECSNumber 236-689-1
RTECSNumber NL1925000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only